Cadmium(II) chloride, anhydrous ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium chloride

AC31527-0050 AC31527-1000 AC31527-5000
TS31527-0050EA 24 USD
TS31527-0050 TS31527-1000 TS31527-5000
Cadmium(II) chloride, anhydrous ACS
Cadmium(II) chloride
Formula: CdCl₂
MW: 183.32 g/mol
Boiling Pt: 960 °C (760 mmHg)
Melting Pt: 568 °C
Storage Temperature: Ambient
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 12,01653

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥99.0 %
Nitrate and Nitrite (as NO3) ≤0.003 %
Sulfate (SO4) ≤0.01 %
Ammonium (NH4) ≤0.01 %
Calcium (Ca) ≤0.01 %
Copper (Cu) ≤0.001 %
Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.05 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR