Manganese(II) chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

AC27141-0010 AC27141-0025 AC27141-0250 AC27141-2500
TS27141-0010EA 56 USD
TS27141-0010 TS27141-0025 TS27141-0250 TS27141-2500
Manganese(II) chloride 97%
Manganese(II) chloride
Formula: MnCl₂
Boiling Pt: 1190 °C (760 mmHg)
Melting Pt: 650 °C
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 12,05768

Order Now

Specification Test Results

Appearance Pink Flakes or beads
Titration Complexometric ≥96.0 %
Water ≤1 %
Iron (Fe) ≤200 ppm
Nickel (Ni) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR