Gold(I) chloride 99+%
Supplier: Thermo Scientific Chemicals
AC38833-0010
AC38833-0050
TS38833-0010EA
278
USD
TS38833-0010
TS38833-0050
Gold(I) chloride 99+%
Gold(I) chloride
Formula:
AuCl MW: 232.42 g/mol Melting Pt: 289 °C |
MDL Number:
MFCD00046175 CAS Number: 10294-29-8 EINECS: 233-655-8 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | Yellow-green Powder |
Elemental Analysis | ≥99.0 % (ex Au) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...