Lead(II) nitrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

AC21156-0010 AC21156-0051 AC21156-5000
TS21156-0010EA 134 USD
TS21156-0010 TS21156-0051 TS21156-5000
Lead(II) nitrate 99+%
Lead(II) nitrate
Formula: Pb(NO₃)₂
Melting Pt: 470 °C (dec.)
Density: 4.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤10 ppm
Iron (Fe) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR