Cerium trichloride heptahydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate

AC21182-0250 AC21182-1000 AC21182-5000
TS21182-0250EA 39 USD
TS21182-0250 TS21182-1000 TS21182-5000
Cerium trichloride heptahydrate 99%
Cerium trichloride heptahydrate
Formula: CeCl₃·7H₂O
MW: 372.58 g/mol
Melting Pt: 848
Storage Temperature: Ambient
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 232-227-8
UN: 1759
ADR: 8 II
Merck Index: 12,02041

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR