Iron(II) bromide, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

AC38872-0100 AC38872-0500
TS38872-0100EA 87 USD
TS38872-0100 TS38872-0500
Iron(II) bromide, anhydrous 98%
Iron(II) bromide
Formula: Br₂Fe
MW: 215.66 g/mol
Melting Pt: 684 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to orange to dark brown Powder and/or lumps
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR