Ammonium bromide 98%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

AC45065-0010 AC45065-0025 AC45065-0100
TS45065-0010EA 64 USD
TS45065-0010 TS45065-0025 TS45065-0100
Ammonium bromide 98%, extra pure
Ammonium bromide
Formula: H₄BrN
MW: 97.94 g/mol
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 542 (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
Merck Index: 13,00505

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥97.5 %
Sulfated ash ≤0.1 %
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR