Aluminium bromide, anhydrous 98+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum bromide

AC19841-0250 AC19841-1000 AC19841-5000
TS19841-0250EA 58 USD
TS19841-0250 TS19841-1000 TS19841-5000
Aluminium bromide, anhydrous 98+%, Extra Pure
Aluminium bromide
Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265 °C (760 mmHg)
Melting Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003421
CAS Number: 7727-15-3
UN: 1725
ADR: 8 II
Merck Index: 13,00330

Order Now

Specification Test Results

Appearance White to yellow to pink to gray Crystalline powder or chunks
Titration Complexometric ≥98.0 %
Chloride (Cl) ≤2000 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR