Cesium bromide 99.9%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC19206-0250 AC19206-1000
TS19206-0250EA 80.8 USD
TS19206-0250 TS19206-1000
Cesium bromide 99.9%, pure
Cesium bromide
Formula: BrCs
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 636 °C
Density: 4.44 g/cm³
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 13,02019

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR