Potassium tetrahydroborate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

AC19655-0025 AC19655-0050 AC19655-1000 AC19655-5000
TS19655-0025EA 989 USD
TS19655-0025 TS19655-0050 TS19655-1000 TS19655-5000
Potassium tetrahydroborate 98+%
Potassium tetrahydroborate
Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 12,07777

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR