Sodium tetraethylborate 97%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC36388-0010
AC36388-0050
TS36388-0010EA
77
USD
TS36388-0010
TS36388-0050
Sodium tetraethylborate 97%, pure
Sodium tetraethylborate
Formula:
C₈H₂₀BNa Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061547 CAS Number: 15523-24-7 UN: 3393 ADR: 4.2, |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...