Osmium tetraoxide 99.9+% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
AC19118-0010
AC19118-0050
AC19118-1000
AC19118-2500
AC19118-5000
TS19118-0010EA
288
USD
TS19118-0010
TS19118-0050
TS19118-1000
TS19118-2500
TS19118-5000
Osmium tetraoxide 99.9+% (trace metals basis)
Osmium tetraoxide
Formula:
O₄Os Boiling Pt: 130 °C (760 mmHg) Melting Pt: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011150 CAS Number: 20816-12-0 EINECS: 244-058-7 UN: 2471 ADR: 6.1,I Merck Index: 13,06962 |
Specification Test Results
Appearance | Yellow Crystals |
Melting point | 40 to 42°C |
Heavy metals | ≤0.005 % |
Solubility | Passes test |
Purity | ≥99.90 % (Metal purity) |
Non-volatile matter | ≤0.2 % |
Osmium (Os) | 74.2 to 75.4 % |
Ruthenium (Ru) | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...