Zirconium tetrafluoride 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconium (IV) fluoride

AC29979-0250 AC29979-1000
TS29979-0250EA 82 USD
TS29979-0250 TS29979-1000
Zirconium tetrafluoride 99.5%
Zirconium tetrafluoride
Formula: F₄Zr
MW: 167.22 g/mol
Boiling Pt: 905 °C
Melting Pt: 640 °C
MDL Number: MFCD00011307
CAS Number: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8 II
Merck Index: 12,10308

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR