Zirconium tetrachloride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconium (IV) chloride

AC20641-0025 AC20641-0050 AC20641-1000 AC20641-5000
TS20641-0025EA 982 USD
TS20641-0025 TS20641-0050 TS20641-1000 TS20641-5000
Zirconium tetrachloride, anhydrous 98%
Zirconium tetrachloride
Formula: Cl₄Zr
MW: 233.03 g/mol
Melting Pt: 437 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8,III
Merck Index: 12,10307

Order Now

Specification Test Results

Appearance White Powder
Assay ≥97.5 % (ZrCl4)
Aluminium (Al) ≤100 ppm
Chromium (Cr) ≤100 ppm
Hafnium (Hf) ≤100 ppm
Iron (Fe) ≤1000 ppm
Silicon (Si) ≤150 ppm
Titanium (Ti) ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR