Silver fluoride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride

AC21045-0050 AC21045-0250
TS21045-0050EA 103 USD
TS21045-0050 TS21045-0250
Silver fluoride 99+%
Silver fluoride
Formula: AgF
MW: 126.87 g/mol
Boiling Pt: 1150
Melting Pt: 433
Density: 5.85 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8 II
Merck Index: 12,08657

Order Now

Specification Test Results

Appearance Brown-yellow to rust brown Powder
Assay ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR