Lanthanum(III) oxide 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
AC19329-0010
AC19329-1000
AC19329-5000
TS19329-0010EA
461
USD
TS19329-0010
TS19329-1000
TS19329-5000
Lanthanum(III) oxide 99.99% (trace metals basis)
Lanthanum(III) oxide
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Specification Test Results
Appearance | White Powder |
Assay | ≥99.94 % (La2O3) TREO base |
Loss on ignition | ≤2.0 % (1000°C) |
Iron(III)oxide (Fe2O3) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...