Lanthanum(III) oxide 91%
Supplier: Thermo Scientific Chemicals
AC41302-1000
AC41302-2500
TS41302-1000EA
88
USD
TS41302-1000
TS41302-2500
Lanthanum(III) oxide 91%
Lanthanum(III) oxide
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Specification Test Results
Appearance | White to light gray Powder |
Assay | ≥89 % |
Melting point | 170 to 175 °C |
GC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...