Lanthanum(III) chloride 99.9%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC31523-0100
AC31523-0500
AC31523-2500
TS31523-0100EA
113
USD
TS31523-0100
TS31523-0500
TS31523-2500
Lanthanum(III) chloride 99.9%
Lanthanum(III) chloride
Formula:
Cl₃La Boiling Pt: 1000 °C Melting Pt: 860 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011068 CAS Number: 10099-58-8 EINECS: 233-237-5 UN: 1759 ADR: 8,III Merck Index: 13,05379 |
Specification Test Results
Appearance | White to off-white Powder |
Total conc. of imp.as REO | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...