Diphenyl diselenide 99%

Supplier: Thermo Scientific Chemicals
Danger

AC17757-0050 AC17757-0250 AC17757-1000
TS17757-0050EA 68 USD
TS17757-0050 TS17757-0250 TS17757-1000
Diphenyl diselenide 99%
Diphenyl diselenide
Formula: C₁₂H₁₀Se₂
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003001
CAS Number: 1666-13-3
EINECS: 216-780-2
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 63 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR