Tellurium tetrachloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tellurium(IV) chloride

AC19474-0250 AC19474-1000
TS19474-0250EA 102 USD
TS19474-0250 TS19474-1000
Tellurium tetrachloride 99%
Tellurium tetrachloride
Formula: Cl₄Te
MW: 269.41 g/mol
Boiling Pt: 380
Melting Pt: 224
Density: 3.26 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS Number: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8 II
Merck Index: 12,09279

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder
Titration Argentometric ≥98.5 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR