Ammonium nitrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC43696-0250
AC43696-1000
TS43696-0250EA
110
USD
TS43696-0250
TS43696-1000
Ammonium nitrate 99.999% (trace metals basis)
Ammonium nitrate
Formula:
NH₄NO₃ Boiling Pt: 210 °C (760 mmHg) Melting Pt: 169 °C Density: 1.72 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011425 CAS Number: 6484-52-2 UN: 1942 ADR: 5.1,III |
Specification Test Results
Appearance | White Crystalline powder |
Total trace metal impurities | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...