Bismuth(III) chloride 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth(III) chloride, Bismuth trichloride

AC31835-0050 AC31835-0250
TS31835-0050EA 85 USD
TS31835-0050 TS31835-0250
Bismuth(III) chloride 99.999% (trace metals basis)
Bismuth(III) chloride
Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: 447 °C (760 mmHg)
Melting Pt: 230…232 °C
Density: 4.75 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
Merck Index: 13,01261

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR