Thallium(I) chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

AC20888-0250 AC20888-1000
TS20888-0250EA 110 USD
TS20888-0250 TS20888-1000
Thallium(I) chloride 99%
Thallium(I) chloride
Formula: ClTl
Boiling Pt: 720 °C (1013 hPa)
Melting Pt: 430 °C
Density: 7 g/cm³
MDL Number: MFCD00011274
CAS Number: 7791-12-0
EINECS: 232-241-4
UN: 1707
ADR: 6.1,II
Merck Index: 12,09395

Order Now

Specification Test Results

Appearance White to light yellow Powder
Titration Argentometric 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR