Aluminium chloride, anhydrous 99.999% (trace metals basis), powder for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum chloride

AC36481-0100 AC36481-0200 AC36481-1000
TS36481-0100EA 160 USD
TS36481-0100 TS36481-0200 TS36481-1000
Aluminium chloride, anhydrous 99.999% (trace metals basis), powder for analysis
Aluminium chloride
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 194 °C
Density: 2.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.001 %
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR