Barium peroxide 95%

Supplier: Thermo Scientific Chemicals
Danger

AC35041-0025 AC35041-0250 AC35041-5000
TS35041-0025EA 354 USD
TS35041-0025 TS35041-0250 TS35041-5000
Barium peroxide 95%
Barium peroxide
Formula: BaO₂
MW: 169.33 g/mol
Melting Pt: 450
Storage Temperature: Ambient
MDL Number: MFCD00003454
CAS Number: 1304-29-6
EINECS: 215-128-4
UN: 1449
ADR: 5.1 II
Merck Index: 12,01018

Order Now

Specification Test Results

Appearance Beige Powder
Titration Iodometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR