Strontium nitrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC31517-0025
AC31517-1000
AC31517-5000
TS31517-0025EA
472
USD
TS31517-0025
TS31517-1000
TS31517-5000
Strontium nitrate 99+% ACS
Strontium nitrate
Formula:
Sr(NO₃)₂ MW: 211.63 g/mol Boiling Pt: 645 Melting Pt: 570 Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1 III Merck Index: 13,08926 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | ≥99.0 % |
Loss on drying | ≤0.1 % (105°C) |
pH | 5.0 to 7.0 (5 % solution at 25.0°C) |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤50 ppm |
Barium (Ba) | ≤500 ppm |
Calcium (Ca) | ≤500 ppm |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤1000 ppm |
Sodium (Na) | ≤1000 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...