Strontium nitrate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC22256-0010
AC22256-2500
TS22256-0010EA
210
USD
TS22256-0010
TS22256-2500
Strontium nitrate 99+% for analysis
Strontium nitrate
Formula:
Sr(NO₃)₂ MW: 211.63 g/mol Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 13,08926 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | ≥99.0 % |
Loss on drying | ≤0.1 % (105°C) |
pH | 5 to 7 (5 % at 20°C) |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤50 ppm |
Barium (Ba) | ≤100 ppm |
Copper (Cu) | ≤2 ppm |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤20 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤50 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...