Barium nitrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC20315-0025 AC20315-5000
TS20315-0025EA 456 USD
TS20315-0025 TS20315-5000
Barium nitrate 99+% for analysis
Barium nitrate
Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: 592
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 13,00986

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥99.0 %
pH 5 to 8 (5 % at 20°C)
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Strontium (Sr) ≤1000 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR