Sodium metavanadate 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium vanadium trioxide

AC26921-0010 AC26921-2500
TS26921-0010EA 410 USD
TS26921-0010 TS26921-2500
Sodium metavanadate 96%
Sodium metavanadate
Formula: NaO₃V
MW: 121.93 g/mol
Melting Pt: 630 °C
Storage Temperature: Ambient
MDL Number: MFCD00011125
CAS Number: 13718-26-8
EINECS: 237-272-7
UN: 3285
ADR: 6.1,II
Merck Index: 12,08849

Order Now

Specification Test Results

Appearance White to yellow to light beige to light green Powder
Titration with Na2S2O3 ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR