Selenious acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19887-0010
AC19887-0500
AC19887-2500
TS19887-0010EA
796
USD
TS19887-0010
TS19887-0500
TS19887-2500
Selenious acid 98%
Selenious acid
Formula:
H₂O₃Se Melting Pt: 70 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011331 CAS Number: 7783-00-8 EINECS: 231-974-7 UN: 3283 ADR: 6.1,II Merck Index: 14,08430 |
Specification Test Results
Appearance | White to off-white to light pink Crystalline powder or crystals and/or chunks |
Titration Iodometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...