Sodium selenite Se 44-46%, anhydrous
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC20073-0250
AC20073-1000
AC20073-5000
TS20073-0250EA
51.1
USD
TS20073-0250
TS20073-1000
TS20073-5000
Sodium selenite Se 44-46%, anhydrous
Sodium selenite
Formula:
Na₂SeO₃ Melting Pt: 350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003489 CAS Number: 10102-18-8 EINECS: 233-267-9 UN: 2630 ADR: 6.1,I Merck Index: 12,08822 |
Specification Test Results
Appearance | White to light beige Powder |
Assay | 44 to 46 % (Se) |
Loss on drying | ≤0.5 % |
Iron (Fe) | ≤300 ppm |
Lead (Pb) | ≤500 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...