Sodium hexafluoroantimonate 98%

Supplier: Thermo Scientific Chemicals
Warning

AC41954-0250 AC41954-1000
TS41954-0250EA 49 USD
TS41954-0250 TS41954-1000
Sodium hexafluoroantimonate 98%
Sodium hexafluoroantimonate
Formula: F₆NaSb
Density: 3.375 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Assay ≥97.5 %
Melting point 80 to 84 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR