Sodium hydrogen sulfate 92% tech., granular
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21485-0010
AC21485-0025
AC21485-0250
TS21485-0010EA
71
USD
TS21485-0010
TS21485-0025
TS21485-0250
Sodium hydrogen sulfate 92% tech., granular
Sodium hydrogen sulphate
Formula:
HNaO₄S MW: 120.06 g/mol Melting Pt: 177…180 °C Density: 2.1 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003531 CAS Number: 7681-38-1 EINECS: 231-665-7 Merck Index: 12,08729 |
Specification Test Results
Appearance | White to light yellow Granules |
Titration with NaOH | ≥90 % (≥37.6 % (equivalent H2SO4)) |
Chloride (Cl) | ≤1000 ppm |
Iron (Fe) | ≤200 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...