Sodium sulfite, anhydrous 98+% ACS

Supplier: Thermo Scientific Chemicals

AC42443-0010 AC42443-0025 AC42443-2500
TS42443-0010EA 101 USD
TS42443-0010 TS42443-0025 TS42443-2500
Sodium sulfite, anhydrous 98+% ACS
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodimetric ≥98.0 %
Free acid Passes test
Chloride (Cl) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Titrable free base ≤0.03 meq/g
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR