Sodium metabisulfite for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC22307-0010
AC22307-0025
AC22307-5000
TS22307-0010EA
118
USD
TS22307-0010
TS22307-0025
TS22307-5000
Sodium metabisulfite for analysis
Sodium metabisulphite
Formula:
Na₂S₂O₅ Melting Pt: 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 EINECS: 231-548-0 Merck Index: 12,08731 |
|
Specification Test Results
Appearance | White Crystalline powder |
Titration Iodimetric | ≥58.5 % (SO2) |
Arsenic (As) | ≤1 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...