Trilithium orthophosphate, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium phosphate tribasic, Lithium orthophosphate, Lithium phosphate

AC38978-1000 AC38978-5000
TS38978-1000EA 52 USD
TS38978-1000 TS38978-5000
Trilithium orthophosphate, extra pure
Trilithium orthophosphate
Formula: Li₃O₄P
MW: 115.79
Melting Pt: 1225
Density: 2.41
Storage Temperature: Ambient
MDL Number: MFCD00016187
CAS Number: 10377-52-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HClO4 ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR