Hydrogen peroxide 12% (w/v)

Supplier: Thermo Scientific Chemicals
Danger

AC46457-0010 AC46457-1000
TS46457-0010EA 49 USD
TS46457-0010 TS46457-1000
Hydrogen peroxide 12% (w/v)
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: 100 °C (760 mmHg)
Density: 1.0500 g/ml
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2984
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with KMnO4 11.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR