Potassium nitrite 97%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC45319-0010 AC45319-0050
TS45319-0010EA 97 USD
TS45319-0010 TS45319-0050
Potassium nitrite 97%, Extra Pure
Potassium nitrite
Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 387 °C
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
UN: 1488
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Titration excess KMnO4 96.0 to 104.0 %
Arsenic (As) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 % (10 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR