Potassium nitrite 97% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC22270-0010 AC22270-2500
TS22270-0010EA 136 USD
TS22270-0010 TS22270-2500
Potassium nitrite 97% for analysis
Potassium nitrite
Formula: KNO₂
Melting Pt: 387 °C
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734

Order Now

Specification Test Results

Appearance White to light yellow Crystals
Titration with KMnO4 ≥96.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR