Sodium amide 95%, pure

Supplier: Thermo Scientific Chemicals
Danger

AC19705-0025 AC19705-1000 AC19705-5000
TS19705-0025EA 382 USD
TS19705-0025 TS19705-1000 TS19705-5000
Sodium amide 95%, pure
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
Boiling Pt: 400 °C
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00011117
CAS Number: 7782-92-5
EINECS: 231-971-0
UN: 3131
ADR: 4.3,I
Merck Index: 12,08717

Order Now

Specification Test Results

Appearance White to grayish-white Powder or irregular granules or flakes
Titration with HClO4 ≥94 %
Free Sodium Passes test (A small amount dropped into water, does not catch fire)
Peroxides ≤0.1 % (as NaNH2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR