Sodium amide 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC33924-0250 AC33924-1000
TS33924-0250EA 55 USD
TS33924-0250 TS33924-1000
Sodium amide 99%, Extra Pure
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
Boiling Pt: 400 °C
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00011117
CAS Number: 7782-92-5
EINECS: 231-971-0
UN: 3131
ADR: 4.3,I
Merck Index: 12,08717

Order Now

Specification Test Results

Appearance White to grayish-white Powder or irregular granules or flakes
Titration with HClO4 ≥98.5 %
Free Sodium Passes test (A small amount dropped into water, does not catch fire)
Peroxides ≤0.1 % (as NaNH2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR