Sodium dicyanoamide 97%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC17634-0250 AC17634-1000 AC17634-5000
TS17634-0250EA 56 USD
TS17634-0250 TS17634-1000 TS17634-5000
Sodium dicyanoamide 97%, pure
Sodium dicyanoamide
Formula: C₂N₃Na
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003535
CAS Number: 1934-75-4
EINECS: 217-703-5

Order Now

Specification Test Results

Appearance White to off-white to light pink Powder
Infrared spectrum Conforms
Titration Argentometric ≤2.0 % (NaCl)
Water ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR