Sodium iodide 99.999% (trace metals basis), Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

AC21268-0010 AC21268-0100 AC21268-1000
TS21268-0010EA 22 USD
TS21268-0010 TS21268-0100 TS21268-1000
Sodium iodide 99.999% (trace metals basis), Extra Pure
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300 °C (760 mmHg)
Melting Pt: 661 °C
Density: 3.66 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Appearance White Granular powder
Total trace metal impurities ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR