Sodium fluoride 97%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

AC19127-0010 AC19127-0250 AC19127-5000
TS19127-0010EA 148 USD
TS19127-0010 TS19127-0250 TS19127-5000
Sodium fluoride 97%, extra pure
Sodium fluoride
Formula: NaF
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
UN: 1690
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder or crystals
Titration ≥96.0 %
Chloride (Cl) ≤120 ppm
Sulfate (SO4) ≤5000 ppm
Sodium fluosilicate ≤1 % (Na2SiF6)
Iron (Fe) ≤200 ppm
Heavy metals (as Pb) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR