Potassium fluoride 40% (w/w) on alumina
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC39081-0250
AC39081-1000
TS39081-0250EA
69
USD
TS39081-0250
TS39081-1000
Potassium fluoride 40% (w/w) on alumina
Potassium fluoride
Formula:
KF MW: 58.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 UN: 1812 ADR: 6.1,III |
Specification Test Results
Appearance | White Powder |
Assay | 35 to 45 wt % (Mass balance) |
Water | 3 to 10 % (K.F.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...