Sodium chromate, anhydrous 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC37787-0250 AC37787-1000 AC37787-5000
TS37787-0050EA 25.08 USD
TS37787-0050 TS37787-0250 TS37787-1000 TS37787-5000
Sodium chromate, anhydrous 98+%
Sodium chromate
Formula: CrNa₂O₄
MW: 161.97 g/mol
Melting Pt: 792 °C
Storage Temperature: Ambient
MDL Number: MFCD00003484
CAS Number: 7775-11-3
EINECS: 231-889-5
UN: 3290
ADR: 6.1,II
Merck Index: 12,08745

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Titration Iodometric ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR