Sodium hexanitritocobaltate(III)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium cobaltnitrite, Trisodium hexanitritocobaltate

AC21486-0250 AC21486-1000 AC21486-5000
TS21486-0250EA 75 USD
TS21486-0250 TS21486-1000 TS21486-5000
Sodium hexanitritocobaltate(III)
Sodium hexanitritocobaltate(III)
Formula: Na₃Co(NO₂)₆
MW: 403.94 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003512
CAS Number: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II
Merck Index: 13,08677

Order Now

Specification Test Results

Appearance Orange to orange-brown Powder
Suitability (for K determination) Passes test
Insoluble matter ≤0.02 % (in dilute acetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR