Potassium chlorate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC41819-0025
AC41819-0050
AC41819-5000
TS41819-0025EA
390
USD
TS41819-0025
TS41819-0050
TS41819-5000
Potassium chlorate 99+% ACS
Potassium chlorate
Formula:
KClO₃ Boiling Pt: 400 °C (1013 hPa) Melting Pt: 356…368 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011361 CAS Number: 3811-04-9 EINECS: 223-289-7 UN: 1485 ADR: 5.1,II Merck Index: 13,07703 |
Specification Test Results
Appearance | White Crystalline powder |
Titration | ≥99.0 % |
Chloride (Cl) | ≤10 ppm |
Bromate (BrO3) | ≤150 ppm |
Sulfate (SO4) | Passes test (limit about 0.002%) |
Calcium (Ca) | ≤20 ppm |
Iron (Fe) | ≤3 ppm |
Magnesium (Mg) | ≤20 ppm |
Sodium (Na) | ≤100 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...