Potassium bromate 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC26839-0010
AC26839-2500
TS26839-0010EA
248
USD
TS26839-0010
TS26839-2500
Potassium bromate 99.5% for analysis
Potassium bromate
Formula:
KBrO₃ Melting Pt: 350 °C Density: 3.27 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011359 CAS Number: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1,II Merck Index: 12,07779 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Iodometric | ≥99.5 % |
Loss on drying | ≤0.1 % (105°C) |
pH | 5 to 9 (5 % at 20°C) |
Bromide (Br) | ≤500 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤5 ppm |
Sodium (Na) | ≤100 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Total nitrogen compounds | ≤0.001 % (as N) |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...