Potassium bromide 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

AC42407-0025 AC42407-5000
TS42407-0025EA 381 USD
TS42407-0025 TS42407-5000
Potassium bromide 99+% ACS
Potassium bromide
Formula: KBr
Boiling Pt: 1435 °C (760 mmHg)
Melting Pt: 734 °C
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or solid and/or chunks
Titration Argentimetric ≥99.0 %
pH 5.0 to 8.8 (5% soln. at 25°C)
Chloride (Cl) ≤0.2 %
Iodide (I) ≤0.001 %
Bromate (BrO3) ≤0.001 %
Iodate (IO3) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR